Learn More
Triethylaluminum, 94%, Thermo Scientific Chemicals
Description
Triethylaluminum is used as a co-catalyst in the industrial production of polyethylene and for the production of medium chain alcohols. Used as a catalyst in Ziegler-Natta polymerization process for vinyl, olefin, diene polymerizations and linear oligomerization and cyclization of unsaturated hydrocarbons. It is also used as a catalyst to produce ethylene gas, chain growth of ethylene, longer chain aluminum alkyls, and in plating aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -50°C |
Density | 0.835 g/mL |
Boiling Point | 128°C to 130°C (50 mmHg) |
Flash Point | 18°C (−1°F) |
Odor | Characteristic |
Packaging | Cylinder |
Quantity | 100 g |
UN Number | UN3394 |
Sensitivity | Moisture sensitive |
Solubility Information | Reacts with water. |
Show More |
RUO – Research Use Only