missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023914
Click to view available options
Quantity:
500 g
2500 g
This item is not returnable. View return policy
78-40-0
C6H15O4P
182.16
MFCD00009077
DQWPFSLDHJDLRL-UHFFFAOYSA-N
triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
6535
CHEBI:45927
triethyl phosphate
CCOP(=O)(OCC)OCC
This item is not returnable. View return policy
78-40-0
C6H15O4P
182.16
MFCD00009077
DQWPFSLDHJDLRL-UHFFFAOYSA-N
triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
6535
CHEBI:45927
triethyl phosphate
CCOP(=O)(OCC)OCC

Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

Solubility
Fully miscible in water at 20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.072
Boiling Point 215°C
Flash Point 115°C (240°F)
Odor Mild
Refractive Index 1.405
Quantity 500 g
Merck Index 14,9674
Solubility Information Fully miscible in water at 20°C.
Formula Weight 182.16
Percent Purity ≥98%
Chemical Name or Material Triethyl phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'