missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals

Catalog Number p-203847
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3699-66-9
C9H19O5P
238.22
MFCD00009159
BVSRWCMAJISCTD-UHFFFAOYSA-N
triethyl 2-phosphonopropionate, ethyl 2-diethoxyphosphoryl propanoate, ethyl-2-diethylphosphono propanoate, diethyl 1-carbethoxy ethylphosphonate, propanoic acid, 2-diethoxyphosphinyl-, ethyl ester, diethyl ethoxycarbonylethyl-phosphonate, diethyl 1-ethoxycarbonyl ethanephosphonate, 2-diethoxyphosphinyl propanoic acid ethyl ester, diethyl 1-ethoxycarbonyl ethanephosphonate∼2-phosphonopropionic acid triethyl ester, triethyl2-phosphonopropionate
107155
ethyl 2-diethoxyphosphorylpropanoate
CCOC(=O)C(C)P(=O)(OCC)OCC
This item is not returnable. View return policy
3699-66-9
C9H19O5P
238.22
MFCD00009159
BVSRWCMAJISCTD-UHFFFAOYSA-N
triethyl 2-phosphonopropionate, ethyl 2-diethoxyphosphoryl propanoate, ethyl-2-diethylphosphono propanoate, diethyl 1-carbethoxy ethylphosphonate, propanoic acid, 2-diethoxyphosphinyl-, ethyl ester, diethyl ethoxycarbonylethyl-phosphonate, diethyl 1-ethoxycarbonyl ethanephosphonate, 2-diethoxyphosphinyl propanoic acid ethyl ester, diethyl 1-ethoxycarbonyl ethanephosphonate∼2-phosphonopropionic acid triethyl ester, triethyl2-phosphonopropionate
107155
ethyl 2-diethoxyphosphorylpropanoate
CCOC(=O)C(C)P(=O)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1100g/mL
Boiling Point 143.0°C to 144.0°C (12.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH(CH3)CO2C2H5
Refractive Index 1.4310 to 1.4330
Quantity 10 g
Beilstein 04, I, 573
Specific Gravity 1.11
Formula Weight 238.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl 2-phosphonopropionate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl 2-phosphonopropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'