missing translation for 'onlineSavingsMsg'
Learn More

Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals

Catalog Number 10498320
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
CCO[Si](OCC)(OCC)C=C
This item is not returnable. View return policy
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
CCO[Si](OCC)(OCC)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.9000g/mL
Boiling Point 160°C to 161°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSi(OC2H5)3
Refractive Index 1.3970 to 1.399
Quantity 25 g
Specific Gravity 0.9
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in chloroform
IUPAC Name ethenyltriethoxysilane
Formula Weight 190.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethoxyvinylsilane, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'