missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine hydrochloride, +99%, ACROS Organics™

Brand:  Acros Organics 170055000

Code : 29

Additional Details : CAS Number : 637-39-8 Weight : 0.55000kg

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10285500

  • £38.00 / 500g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Triethanolamine hydrochloride
637-39-8
99+%
(HOCH2CH2)3N·HCl
MFCD00012596
p.a.
≥99.0%
C6H15NO3·HCl
185.65
2, 2', 2''-Nitrilotriethanol hydrochloride

Specifications

Triethanolamine hydrochloride
p.a.
Crystalline Powder
99+%
(HOCH2CH2)3N·HCl
185.65
1.0% max. (105°C)
Glass bottle
500g
637-39-8
≥99.0%
C6H15NO3·HCl
Authentic
MFCD00012596
2, 2', 2''-Nitrilotriethanol hydrochloride
White
Videos
SDS