missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10152640
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
2855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
2855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO

Triethanolamine, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point 21°C
Color Yellow
pH 10.5
Boiling Point 360°C
Packaging Amber glass bottle
Quantity 2.5 L
Vapor Density 5.14
Formula Weight 149.19g/mol
Vapor Pressure <0.01mmHg at 20°C
Physical Form Viscous Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'