missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391303
288.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
C6H15NO3
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
7618
CHEBI:28621
2-[bis(2-hydroxyethyl)amino]ethanol
C(CO)N(CCO)CCO

High quality chemicals for laboratory and specialized industrial use

Flash Point 179.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.13kg
Formula Weight 149.19
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Triethanolamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'