missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine 97%, ACROS Organics™

Brand:  Acros Organics 139560010

Code : 29

Additional Details : CAS Number : 102-71-6 Weight : 1.05000kg

Quantity 1L

 View more versions of this product

Product Code. 10123000

  • £37.20 / 1 L


Estimated Shipment
13-06-2018
Add to basket

Specifications

Chemical Identifiers

Triethanolamine
102-71-6
97%
(HOCH2CH2)3N
MFCD00002855
97%
97%
C6H15NO3
149.19
2, 2', 2''-Nitrilotriethanol, 2, 2';, 2';';-Nitrilotriethanol, TEA

Specifications

Triethanolamine
97%
102-71-6
Oily Liquid
97%
(HOCH2CH2)3N
149.19
01,1196
2, 2', 2''-Nitrilotriethanol, 2, 2';, 2';';-Nitrilotriethanol, TEA
1.4840 to 1.4860
>325°C
21.0°C
190°C
1L
Vapor Pressure: <0.01mmHg at 20°C
1.1245g/mL
97%
C6H15NO3
Authentic
MFCD00002855
15, 9830
Solubility in water: freely soluble. Other solubilities: miscible with alcohol, methanol and acetone, soluble in chloroform, slichtly soluble in benzene and ether, soluble 4.2% in benzene; 1.6% in ether; 0.4% in, carbon tetrachloride and < 0.1% in n-hepta
1.1245
360.0°C
Clear
Videos
SDS