missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog Number p-3523611
63.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
58656-04-5
C18H34BF4P
368.25
MFCD03840580
MYSMMEUXKHJYKH-UHFFFAOYSA-O
tricyclohexylphosphonium tetrafluoroborate, tricyclohexylphosphine tetrafluoroborate, tricyclohexylphosphine tetrafluroborate, tricyclohexylphosphoniumtetrafluoroborate, pcy3.hbf4, ksc269e4n, tricyclohexylphosphine tetrofluroborate, tricyclohexyl phosphonium tetrafluoroborate, tetrafluoroboranuide tricyclohexylphosphanium, tricyclohexyl-phosphonium tetrafluoro borate
11710396
F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
58656-04-5
C18H34BF4P
368.25
MFCD03840580
MYSMMEUXKHJYKH-UHFFFAOYSA-O
tricyclohexylphosphonium tetrafluoroborate, tricyclohexylphosphine tetrafluoroborate, tricyclohexylphosphine tetrafluroborate, tricyclohexylphosphoniumtetrafluoroborate, pcy3.hbf4, ksc269e4n, tricyclohexylphosphine tetrofluroborate, tricyclohexyl phosphonium tetrafluoroborate, tetrafluoroboranuide tricyclohexylphosphanium, tricyclohexyl-phosphonium tetrafluoro borate
11710396
F[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165.0°C to 169.0°C
Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
IUPAC Name tetrafluoroboranuide; tricyclohexylphosphanium
Formula Weight 368.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Tricyclohexylphosphonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'