missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine, technical grade, Thermo Scientific Chemicals

Catalog Number p-7059558
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2622-14-2
C18H33P
280.436
MFCD00003853
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
75806
tricyclohexylphosphane
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
This item is not returnable. View return policy
2622-14-2
C18H33P
280.436
MFCD00003853
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
75806
tricyclohexylphosphane
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 78°C
Color White
Odor Unpleasant
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 280.42
Grade Technical
Physical Form Solid
Chemical Name or Material Tricyclohexylphosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'