missing translation for 'onlineSavingsMsg'
Learn More

Tricyclohexylphosphine oxide, Thermo Scientific Chemicals

Catalog Number p-7049859
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
13689-19-5
C18H33OP
296.44
MFCD00014298
LEFPWWWXFFNJAA-UHFFFAOYSA-N
tricyclohexylphosphine oxide, phosphine oxide, tricyclohexyl, dicyclohexylphosphoroso cyclohexane, tricyclohexylphosphineoxide, acmc-1bo3x, 4-16-00-01009 beilstein handbook reference, tricyclohexylphosphine oxide 1g
26187
dicyclohexylphosphorylcyclohexane
O=P(C1CCCCC1)(C1CCCCC1)C1CCCCC1
This item is not returnable. View return policy
13689-19-5
C18H33OP
296.44
MFCD00014298
LEFPWWWXFFNJAA-UHFFFAOYSA-N
tricyclohexylphosphine oxide, phosphine oxide, tricyclohexyl, dicyclohexylphosphoroso cyclohexane, tricyclohexylphosphineoxide, acmc-1bo3x, 4-16-00-01009 beilstein handbook reference, tricyclohexylphosphine oxide 1g
26187
dicyclohexylphosphorylcyclohexane
O=P(C1CCCCC1)(C1CCCCC1)C1CCCCC1

Tricyclohexylphosphine oxide is used as a ligand for a range of metal-catalyzed organic transformations. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides.1 Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides. This ligand is applied with Ni as a key intermediate in the formation of cylcopentane compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 155°C to 157°C
Quantity 1 g
UN Number UN3464
Solubility Information Soluble in most organic solvents. Insoluble in water.
Formula Weight 296.42
Physical Form Crystalline
Chemical Name or Material Tricyclohexylphosphine oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'