missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, 99%, Thermo Scientific Chemicals

Catalog Number p-7042931
Click to view available options
Quantity:
100 g
500 g
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
N-(Trishydroxymethylmethyl)glycine
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO
5704-04-1
C6H13NO5
179.17
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
N-(Trishydroxymethylmethyl)glycine
79784
CHEBI:39063
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C(C(=O)O)NC(CO)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽183°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula (HOCH2)3CNHCH2CO2H
Beilstein 1937804
Merck Index 14,9651
Formula Weight 179.17
Percent Purity 99%
Grade Electrophoresis
Chemical Name or Material Tricine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'