missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tricine, 99+%, for biochemistry
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | (0.1 M aq. soln.) at 0nm |
Absorbance Conditions | (0.1M aq. soln.) |
Melting Point | 180.0°C to 184.0°C |
Color | White |
pH | 4.6 to 5.6 (1% aq. soln. at 25°C) |
Physical Form | Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Linear Formula | (HOCH2)3CNHCH2CO2H |
Show More |
Safety and Handling
Product Identifier
- Tricine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'