missing translation for 'onlineSavingsMsg'
Learn More

Trichlorovinylsilane, 96%, Thermo Scientific™

Catalog Number 10675192
Click to view available options
10mL; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 75-94-5
Density 1.2700g/mL
Boiling Point 90.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C2H3Cl3Si
Linear Formula H2C=CHSiCl3
Refractive Index 1.4350 to 1.4370
MDL Number MFCD00000482
Quantity 10mL
Fieser 05,749
Specific Gravity 1.27
Synonym trichlorovinylsilane, vinyltrichlorosilane, trichloro vinyl silane, vtcs, silane, trichloroethenyl, silane, trichlorovinyl, trichlorovinylsilicon, vinyl trichlorosilane, trichlorovinyl silicane, vinylsilicon trichloride
Solubility Information Solubility in water: reacts
InChI Key GQIUQDDJKHLHTB-UHFFFAOYSA-N
SMILES C=C[Si](Cl)(Cl)Cl
IUPAC Name trichloro(ethenyl)silane
Molecular Weight (g/mol) 161.49
PubChem CID 6413
Viscosity 0.6 mPa.s (20°C)
Formula Weight 161.49
Percent Purity 96%
Chemical Name or Material Trichlorovinylsilane, 96%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trichlorovinylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'