missing translation for 'onlineSavingsMsg'
Learn More

α,α,α-Trichlorotoluene, 98%, Thermo Scientific Chemicals

Catalog Number p-202903
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
98-07-7
C7H5Cl3
195.47
MFCD00000786
XEMRAKSQROQPBR-UHFFFAOYSA-N
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
7367
CHEBI:82274
trichloromethylbenzene
ClC(Cl)(Cl)C1=CC=CC=C1
This item is not returnable. View return policy
98-07-7
C7H5Cl3
195.47
MFCD00000786
XEMRAKSQROQPBR-UHFFFAOYSA-N
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
7367
CHEBI:82274
trichloromethylbenzene
ClC(Cl)(Cl)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -7.5°C
Color Yellow
Density 1.3800g/mL
Boiling Point 219°C to 223°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CCl3
Refractive Index 1.5570 to 1.559
Quantity 5 mL
Beilstein 05, 300
Merck Index 15, 1112
Specific Gravity 1.38
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in alcohol,benzene,ether and,most organic solvents
Formula Weight 195.48
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α, α, α-Trichlorotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trichlorotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'