Trichloroethylene, 99+%, extra pure, stabilized, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 158310250

25LT Trichloroethylene, 99+%, extra pure, stabilized

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 79-01-6 Weight: 36.55000kg Transport: UN number : 1710 Chem class : 1 Pack group : III


Packaging Metal drum
Quantity 25L

Product Code. 10034093

Quantity Price
1 £ 339.3 / 25 L
EU Stock 5
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Trichloroethylene
Name Note Stabilized
CAS 79-01-6
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C2HCl3
Linear Formula ClCH=CCl2
Formula Weight 131.39
MDL Number MFCD00000838

Specification

Assay ≥99.0%
Purity Grade Extra Pure
Physical Form Liquid
Quantity 25L
Color Clear
Linear Formula ClCH=CCl2
Additional Information Vapor Pressure: 77.3mbar at 20°C
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 79-01-6
Assay Percent Range 99+%
MDL Number MFCD00000838
Refractive Index 1.4760 to 1.4780 (20°C, 589nm)
Fieser 09,479; 10,414; 11,552
Packaging Metal drum
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
Water 0.006% max. (Coulometric)
Name Note Stabilized
Density 1.4600g/mL
Molecular Formula C2HCl3
Formula Weight 131.39
Health Hazard 1 Danger
Chemical Name or Material Trichloroethylene
Non_volatile Matter 0.002% max.
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.