missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, 99+%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog Number p-3737642
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
79-01-6
MFCD00000838
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
79-01-6
MFCD00000838
XSTXAVWGXDQKEL-UHFFFAOYSA-N
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
6575
CHEBI:16602
1,1,2-trichloroethene
C(=C(Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -85°C
Density 1.4600g/mL
Boiling Point 87.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Molecular Formula C2HCl3
Linear Formula ClCH=CCl2
Refractive Index 1.4760 to 1.4780
Quantity 2.5 L
Beilstein 01,187
Fieser 09,479; 10,414; 11,552
Merck Index 15,9804
Specific Gravity 1.46
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents
Molecular Weight (g/mol) 131.39
Viscosity 0.55 mPa.s (25°C)
Formula Weight 131.39
Vapor Pressure 77.3mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Trichloroethylene, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trichloroethylene, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'