Trichloroethylene, 99.6%, ACS reagent, stabilized, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 421520025

2.5LT Trichloroethylene, 99.6%, ACS reagent, stabilized

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 79-01-6 Weight: 5.15000kg Transport: UN number : 1710 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 2.5L

Product Code. 10646794

Quantity Price
1 £ 58.4 / 2.50 L
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Trichloroethylene
CAS 79-01-6
Purity Grade ACS Reagent Grade
Assay ≥99.5%
Assay Percent Range 99.6%
Molecular Formula C2HCl3
Linear Formula ClCH=CCl2
Formula Weight 131.39
MDL Number MFCD00000838

Specification

Assay ≥99.5%
Purity Grade ACS Reagent Grade
Physical Form Liquid
Quantity 2.5L
Color Clear
EINECS Number 201-167-4
Heavy Metals (as Pb) 1ppm max.
Linear Formula ClCH=CCl2
Residue after Evaporation 0.001% max.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 79-01-6
Assay Percent Range 99.6%
MDL Number MFCD00000838
Stabilizer 0.5% (typically) Ethanol, or 0.8% (typically) Ethyloxirane
RTECS Number KX4550000
Fieser 09,479; 10,414; 11,552
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
Water 0.02% max. (Coulometric)
Molecular Formula C2HCl3
Formula Weight 131.39
Health Hazard 1 Danger
Titratable Base 0.0003 meq/g max.
Density 1.4600g/mL
Chemical Name or Material Trichloroethylene
TSCA TSCA
Merck Index 15, 9804
Free Halogens Passes Test
Beilstein 01, 187
Titratable Acid 0.0001 meq/g max.
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement:
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.