missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trichloroacrylic acid, 98%

Brand:  Alfa Aesar™ A12299.06

Code : NEW

Additional Details : CAS Number : 2257-35-4 Weight : 0.00500kg Transport : UN number : 3261 Chem class : 1a Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11485033

  • £59.47 / 5g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

Trichloroacrylic acid
C3HCl3O2
MFCD00015455
1754536
2257-35-4
175.4
UN3261
Trichloropropenoic acid

Specifications

Trichloroacrylic acid
98%
175.4
UN3261
Trichloropropenoic acid
5g
2257-35-4
C3HCl3O2
MFCD00015455
1754536
72° to 74°C
Videos
SDS