missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical T/2950/50

Code : 21

Additional Details : CAS Number : 76-03-9 Weight : 0.25000kg Transport : UN number : 1839 Chem class : 8 Pack group : II


Quantity 250g

 Lernen Sie weitere Varianten dieses

Artikelnummer. 10050450

Menge Preis
1 £45.57 / 250g
4 £41.42 / 250g
16 £39.35 / 250g
In stock, ready to ship: 15

Please to purchase this item. Need a web account? Register with us today!



Beschreibung

Beschreibung

Trichloroacetic Acid, Extra Pure, SLR

Spezifikation

Chemische Identifikatoren

76-03-9
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
6421
2,2,2-trichloroacetic acid
C2HCl3O2
4177
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
CHEBI:30956
C(=O)(C(Cl)(Cl)Cl)O

Spezifikation

76-03-9
4177
YNJBWRMUSHSURL-UHFFFAOYSA-N
2,2,2-trichloroacetic acid
6421
163.39g/mol
Plastic powder jar
52°C
White
250g
C2HCl3O2
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloracetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
C(=O)(C(Cl)(Cl)Cl)O
163.378
CHEBI:30956
Solid
1.2mbar at 50°C, 0.08mbar @25C
1.2
196°C
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

Produktidentifikator
  • Trichloroacetic acid

Signalwort
  • Danger

Gefahrenkategorie
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Gefahrenhinweis
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Sicherheitsdatenblatt (SDS) Dokumentation
Zertifikate