missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic acid, 99%, extra pure, Thermo Scientific Chemicals
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
27.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
27.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 52.0°C to 58.0°C |
Boiling Point | 196.0°C |
Color | White |
Assay Percent Range | 0.99 |
Linear Formula | CCl3CO2H |
Beilstein | 2206 |
Fieser | 01,1194; 02,425; 04,520; 15,168 |
Merck Index | 159792 |
Solubility Information | Solubility in water: 120g/100 mL (20°C). Other solubilities: soluble in alcohol and ether,carbon tetrachloride |
Formula Weight | 163.39 |
Show More |
Safety and Handling
Product Identifier
- Trichloroacetic acid
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'