missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trichloro(indenyl)titanium(IV), 98%

Brand:  Alfa Aesar™ 043178.06

Code : NEW

Additional Details : CAS Number : 84365-55-9 Weight : 0.00500kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11314868

  • £260.12 / 5g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Trichloro (indenyl)titanium (IV)
C9H7TiCl3
MFCD00269849
(Indenyl)titanium(IV) trichloride
84365-55-9
269.4
UN3261

Specifications

Trichloro (indenyl)titanium (IV)
Crystalline
C9H7TiCl3
MFCD00269849
(Indenyl)titanium(IV) trichloride
5g
84365-55-9
98%
269.4
UN3261
Purple
Air & Moisture Sensitive
Videos
SDS