missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(vinyl)tin, 97%, ACROS Organics™

£27.22 - £287.08

Chemical Identifiers

CAS 7486-35-3
Molecular Formula C14H30Sn
Molecular Weight (g/mol) 317.09
MDL Number MFCD00009421
InChI Key QIWRFOJWQSSRJZ-UHFFFAOYSA-N
Synonym tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
PubChem CID 81998
IUPAC Name tributyl(ethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10081761
SDS View Documents
Acros Organics
350000010
1g Glass bottle £27.22
1g View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10245771
SDS View Documents
Acros Organics
350000050
5g Glass bottle £93.72
5g View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
10104483
SDS View Documents
Acros Organics
350000250
25g Glass bottle £287.08
25g View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

7486-35-3
317.09
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
81998
CCCC[Sn](CCCC)(CCCC)C=C
C14H30Sn
MFCD00009421
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
tributyl(ethenyl)stannane

Specifications

Tributyl(vinyl)tin
>110°C
Glass bottle
1.08
Colorless to Yellow
7486-35-3
C14H30Sn
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl(ethenyl)stannane
81998
Liquid
1.0800g/mL
Authentic
1.4770 to 1.479
104°C to 106°C (3.5mmHg)
1g
96% min. (GC)
MFCD00009421
Solubility in water: .. Other solubilities: soluble in all the common organic solvents
CCCC[Sn](CCCC)(CCCC)C=C
317.09
317.09
97%
Safety and Handling

Safety and Handling

Product Identifier
  • Tributyl(vinyl)tin

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.