missing translation for 'onlineSavingsMsg'
Learn More

Tributyl(vinyl)tin, 97%, Thermo Scientific Chemicals

Catalog Number 10104483
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7486-35-3
C14H30Sn
317.09
MFCD00009421
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
81998
tributyl(ethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C=C
This item is not returnable. View return policy
7486-35-3
C14H30Sn
317.09
MFCD00009421
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
81998
tributyl(ethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 104°C to 106°C (3.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.479
Quantity 25 g
Specific Gravity 1.08
Solubility Information Solubility in water: .. Other solubilities: soluble in all the common organic solvents
Formula Weight 317.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tributyl(vinyl)tin
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tributyl(vinyl)tin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'