Learn More
Thermo Scientific Maybridge Tributyl(thiazol-5-yl)stannane, 1g, 97%
Brand: Thermo Scientific Maybridge MO08371DA
Tributyl(thiazol-5-yl)stannane, 1g, 97%
Code : 31
Additional Details : CAS Number : 157025-33-7 Weight : 0.05100kg Transport : UN number : 2788 Chem class : 6.1 Pack group : III