missing translation for 'onlineSavingsMsg'
Learn More

Tributyl citrate, 99+%, Thermo Scientific Chemicals

Catalog Number p-3737611
26.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
77-94-1
C18H32O7
360.45
MFCD00027217
ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
6507
tributyl 2-hydroxypropane-1,2,3-tricarboxylate
CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
77-94-1
C18H32O7
360.45
MFCD00027217
ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
6507
tributyl 2-hydroxypropane-1,2,3-tricarboxylate
CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.02% max. (as citric acid)
Melting Point -20°C
Density 1.0420g/mL
Boiling Point 325°C
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOCCOO(CH2)3CH3(CH2COO(CH2)3CH3)2
Refractive Index 1.4430 to 1.446
Quantity 250 g
Merck Index 15, 1565
Specific Gravity 1.042
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic liquids,soluble in ethanol and ether
Viscosity 31.9 mPa.s (25°C)
Formula Weight 360.45
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Tributyl citrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'