missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tributyl(4-methyl-1,3-thiazol-2-yl)stannane, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 388.18 |
Percent Purity | 90% |
Chemical Name or Material | tributyl(4-methyl-1, 3-thiazol-2-yl)stannane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'