missing translation for 'onlineSavingsMsg'
Learn More

Tribromoacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-202391
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
75-96-7
C2HBr3O2
296.73
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid, acetic acid, tribromo, tribromacetic acid, unii-71ktl99qjx, ccris 7887, 71ktl99qjx, acetic acid, 2,2,2-tribromo, snpljqbkdvidtaetp@, dsstox_cid_1668, acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O
This item is not returnable. View return policy
75-96-7
C2HBr3O2
296.73
MFCD00004167
QIONYIKHPASLHO-UHFFFAOYSA-N
tribromoacetic acid, acetic acid, tribromo, tribromacetic acid, unii-71ktl99qjx, ccris 7887, 71ktl99qjx, acetic acid, 2,2,2-tribromo, snpljqbkdvidtaetp@, dsstox_cid_1668, acmc-209p0y
6415
2,2,2-tribromoacetic acid
C(=O)(C(Br)(Br)Br)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 133°C
Color White to Yellow
Boiling Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CBr3CO2H
Quantity 5 g
Beilstein 02, 220
Merck Index 15, 9770
Formula Weight 296.73
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Tribromoacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Tribromoacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'