missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Triangular PTFE Stir Bars

Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container. The triangular PTFE stir bar provides stable stirring action for flat-bottom containers.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.01316kg

 View more versions of this product

Product Code. 11587812

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Stable, general-purpose magnetic stir bar
  • Particularly effective for dissolving solids and mixing sediments due to scraping-like action on the base of the container
  • Autoclavable
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
Containers & Vessels / Stirrer Hot Plates
5/Pk.
8mm
Triangular
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
25 x 8mm
8mm
Stir bar
25mm
Videos
Documentation