missing translation for 'onlineSavingsMsg'
Learn More

Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%, Stabilized, Thermo Scientific Chemicals

Catalog Number p-202352
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C
This item is not returnable. View return policy
1025-15-6
C12H15N3O3
249.27
MFCD00006554
KOMNUTZXSVSERR-UHFFFAOYSA-N
triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
13931
1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 24.8°C
Color Colorless to Yellow
Density 1.1590g/mL
Boiling Point 149.0°C to 152.0°C (4.0 mmHg)
Flash Point 245°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5120 to 1.5140
Quantity 100 g
Beilstein 26, IV, 637
Specific Gravity 1.159
Solubility Information Solubility in water: >1g/L (20°C). Other solubilities: soluble in acetone,dimethyl sulfoxide,ethanol
Stabilizer 1200ppm MEHQ
Viscosity 90.1 mPa.s (30°C)
Formula Weight 249.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triallyl-s-triazine-2,4,6(1H,3H,5H)-trione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'