missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Sodium Citrate Dihydrate, for Electrophoresis, Fisher Chemical™
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Tri-Sodium Citrate Dihydrate, for Electrophoresis
Specifications
Melting Point | >300°C |
Color | White |
pH | 8.4 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 294.09g/mol |
Packaging | Plastic keg |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'