Learn More
Thermo Scientific Alfa Aesar Tri(o-tolyl) phosphite, typically C 71%, H 6%
Brand: Thermo Scientific Alfa Aesar 010258.14
Tri(o-tolyl) phosphite, typically C 71%, H 6%
Code : NEW
Additional Details : CAS Number : 2622-08-4 Weight : 0.02500kg