missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals

Catalog Number p-2692406
29.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-30-8
C21H21O4P
368.37
MFCD00041906
YSMRWXYRXBRSND-UHFFFAOYSA-N
tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
6527
tris(2-methylphenyl) phosphate
CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-30-8
C21H21O4P
368.37
MFCD00041906
YSMRWXYRXBRSND-UHFFFAOYSA-N
tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
6527
tris(2-methylphenyl) phosphate
CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -25°C
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 410°C
Flash Point 225°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.561
Quantity 25 g
Merck Index 15, 9941
Specific Gravity 1.2
Solubility Information Solubility in water: 0.01g/100mL (25°C). Other solubilities: slightly soluble in alcohol,soluble in ether
Formula Weight 368.36
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material Tri-o-tolyl phosphate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-o-tolyl phosphate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H370-Causes damage to organs.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'