missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-o-tolyl phosphate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -25°C |
Color | Colorless to Yellow |
Density | 1.2000g/mL |
Boiling Point | 410°C |
Flash Point | 225°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5580 to 1.561 |
Quantity | 100 g |
Show More |
Safety and Handling
Product Identifier
- Tri-o-tolyl phosphate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after single exposure Category 1
- H411-Toxic to aquatic life with long lasting effects.
- H370-Causes damage to organs.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'