missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-acetyl-D-glucal, 99%, ACROS Organics™

£53.61 - £166.24

Chemical Identifiers

CAS 2873-29-2
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00063253
InChI Key LLPWGHLVUPBSLP-UTUOFQBUSA-N
Synonym tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
PubChem CID 688303
IUPAC Name [(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051500
SDS View Documents
Acros Organics
139240250
25g Glass bottle
£53.61
25g
View alternative products
Save Up to 
»
Estimated Shipment
12-11-2020
Please to purchase this item. Need a web account? Register with us today!
10255380
SDS View Documents
Acros Organics
139241000
100g Glass bottle
£166.24
100g
View alternative products
Save Up to 
»
Estimated Shipment
11-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

2873-29-2
272.25
LLPWGHLVUPBSLP-UTUOFQBUSA-N
688303
CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C
C12H16O7
MFCD00063253
tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
[(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate

Specifications

Tri-O-acetyl-D-glucal
Authentic
Glass bottle
− 15.00
White
25g
2873-29-2
C12H16O7
17, I, 111
Solubility in water: slightly soluble
CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C
272.25
272.25
99%
>110°C
0.5% max. (30°C, 15 hrs) (vacuum)
−15° (20°C c=2,EtOH)
0.1% max.
51.0°C to 53.0°C
99%
98.5% min. (GC)
MFCD00063253
tri-o-acetyl-d-glucal, 3,4,6-tri-o-acetyl-d-glucal, 2r,3s,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-glucal triacetate, 3,4,6-tri-o-acetyl-1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, arabino-hex-1-enitol, 1,5-anhydro-2-deoxy-, triacetate, tri-o-acetylglucal, triacetyl-d-glucal, 2r,3s,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, tri-o-acetyl-glucal
LLPWGHLVUPBSLP-UTUOFQBUSA-N
[(2R,3S,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
688303
Crystalline Powder
SDS