missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyltin chloride, 95%, Thermo Scientific Chemicals

Catalog Number 11304198
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2279-76-7
C9H21ClSn
283.427
MFCD00013610
FVFLIQMADUVDSP-UHFFFAOYSA-M
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
16784
chloro(tripropyl)stannane
CCC[Sn](CCC)(CCC)Cl
This item is not returnable. View return policy
2279-76-7
C9H21ClSn
283.427
MFCD00013610
FVFLIQMADUVDSP-UHFFFAOYSA-M
tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
16784
chloro(tripropyl)stannane
CCC[Sn](CCC)(CCC)Cl

Tri-n-propyltin chloride is used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -23.5°C
Density 1.2678 g/mL at 28°C
Boiling Point 123°C (13 mmHg)
Refractive Index 1.491
Quantity 1 g
UN Number UN2788
Sensitivity Moisture sensitive
Solubility Information Sparingly Soluble in water (4.0E-3g/L) (25°C).
Formula Weight 283.41
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-propyltin chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'