missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tri-n-butyltin iodide, tech. 90%, stab. with copper

Brand:  Alfa Aesar™ H55911.14

Code : NEW

Additional Details : CAS Number : 7342-47-4 Weight : 0.02500kg Transport : UN number : 2788 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15462346

  • £87.00 / 25g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Tri-n-butyltin iodide
Stabilized with copper
[CH3(CH2)3]3SnI
MFCD00074996
7342-47-4
Technical Grade
416.94
UN2788

Specifications

Tri-n-butyltin iodide
Light Sensitive
Colorless
Stabilized with copper
1.5300
Liquid
[CH3(CH2)3]3SnI
MFCD00074996
1.46g/mL
172°C (10mmHg)
25g
7342-47-4
Technical Grade
90%
416.94
UN2788
Videos
SDS