missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tri-n-butylphosphine, 94%

Brand:  Alfa Aesar™ 014114.95

Code : NEW

Additional Details : CAS Number : 998-40-3 Weight : 0.01000kg Transport : UN number : 3254 Chem class : 1 Pack group : I

Quantity 2 x 500g

 View more versions of this product

Product Code. 15491057

  • £438.80 / Each


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Description

Description

  • For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels
Specifications

Chemical Identifiers

Tri-n-butylphosphine
202.32
UN3254
998-40-3
MFCD00009462
1738261

Specifications

Tri-n-butylphosphine
>93° (199°F)
240°C
2 x 500g
Unpleasant
Liquid
202.32
UN3254
Insoluble in water; Miscible with ether, methanol, ethanol and benzene
0.820g/mL
Air & Moisture Sensitive
−65°C
998-40-3
1.4620
94%
MFCD00009462
1738261
Videos
SDS