missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylgermanium hydride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7052156
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
998-39-0
C12H27Ge
243.978
MFCD00077987
UMRPCNGKANTZSN-UHFFFAOYSA-N
tri-n-butylgermane, tributylgermylradical, tri-n-butylgermanium, hydride
6327830
tributylgermanium
CCCC[Ge](CCCC)CCCC
This item is not returnable. View return policy
998-39-0
C12H27Ge
243.978
MFCD00077987
UMRPCNGKANTZSN-UHFFFAOYSA-N
tri-n-butylgermane, tributylgermylradical, tri-n-butylgermanium, hydride
6327830
tributylgermanium
CCCC[Ge](CCCC)CCCC

Reduction of benzylic chlorides

Tri-n-butylgermanium hydride is used in the reduction of benzylic chlorides. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.92 g/mL
Boiling Point 123°C (20 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.4508
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Not miscible.
Formula Weight 244.95
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Tri-n-butylgermanium hydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'