missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tri-n-butylborane, 98%

Brand:  Alfa Aesar™ 033533.06

Code : NEW

Additional Details : CAS Number : 122-56-5 Weight : 0.00500kg Transport : UN number : 2845 Chem class : 1 Pack group : I

Quantity 5g

 View more versions of this product

Product Code. 15461327

  • £21.20 / 5g


Estimated Shipment date
from Supplier 23-03-2018
Add to basket

Specifications

Chemical Identifiers

Tri-n-butylborane
182.16
UN2845
122-56-5
MFCD00009423
Tri-n-butylboron

Specifications

Tri-n-butylborane
−20° (−4°F)
170°C (222mmHg to 225mmHg)
5g
1.4285
98%
MFCD00009423
Tri-n-butylboron
0.747g/mL
Air Sensitive
−34°C
122-56-5
Liquid
182.16
UN2845
Videos
SDS