missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals

Catalog Number p-7023040
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
This item is not returnable. View return policy
126-73-8
C12H27O4P
266.32
MFCD00009436
STCOOQWBFONSKY-UHFFFAOYSA-N
tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
31357
CHEBI:35019
tributyl phosphate
CCCCOP(=O)(OCCCC)OCCCC
This item is not returnable. View return policy
126-73-8
C12H27O4P
266.32
MFCD00009436
STCOOQWBFONSKY-UHFFFAOYSA-N
tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
31357
CHEBI:35019
tributyl phosphate
CCCCOP(=O)(OCCCC)OCCCC

Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

Solubility
Miscible with diethyl ether, benzene, carbon disulfide and ethanol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali.
TRUSTED_SUSTAINABILITY
Melting Point -80°C
Density 0.977
Boiling Point 155°C to 156°C (10 mmHg)
Flash Point 152°C (305°F)
Assay Percent Range 98%
Odor Odorless
Refractive Index 1.424
Quantity 250 mL
Beilstein 1710584
Merck Index 14,9618
Solubility Information Miscible with diethyl ether,benzene,carbon disulfide and ethanol. Slightly miscible with water.
Formula Weight 266.32
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-n-butyl phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'