missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(m-tolyl)phosphine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
Tri(m-tolyl)phosphine is used in suzuki reaction. It acts as a ligand which is used in organometallic chemistry.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store away from incompatible materials such as oxidizing agents.
Specifications
Melting Point | 99°C to 101°C |
Quantity | 1 g |
Beilstein | 658863 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 304.37 |
Percent Purity | ≥98% |
Chemical Name or Material | Tri(m-tolyl)phosphine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'