missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Tri-Cornered Polypropylene Beakers

Catalog Number 11749398
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
400 mL
800 mL
For laboratory use only
For laboratory use only

Tri-cornered lip for dripless pouring

Sturdy enough for autoclaving and reuse, yet inexpensive enough to be disposable

  • Unaffected by commonly used acids, alkalis and solvents
  • Easy-to-read graduations in L and mL
  • Accurate to ±5.0%
  • 10mL graduations
Accuracy ±5%
Capacity (Metric) 50 mL
Material Polypropylene
For Use With (Application) Suitable for Use with Commonly Used Acids, Alkalis and Solvents
Type Beaker
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'