missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-Ammonium Citrate, Certified AR for analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Description
Tri-Ammonium Citrate, Certified AR for analysis
Specifications
Melting Point | 185°C |
Color | White |
pH | 7 to 8 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 243.22g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Citric acid, triammonium salt
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'