missing translation for 'onlineSavingsMsg'
Learn More

Tri(4-morpholinyl)phosphine oxide, 99%, Thermo Scientific Chemicals

Catalog Number p-7046778
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4441-12-7
MFCD00047406
This item is not returnable. View return policy
4441-12-7
MFCD00047406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 188°C to 192°C
Molecular Formula C24H42
Quantity 5 g
Beilstein 280155
Synonym 1-phenyloctadecane, benzene, octadecyl, n-octadecylbenzene, stearylbenzene, 2-phenyloctadecane, octadecane, 1-phenyl, n-octadecyl benzene, acmc-2097fm, octadecane, 1-phenyl-8ci, 1-phenyloctadecane, analytical standard
InChI Key WSVDSBZMYJJMSB-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCC1=CC=CC=C1
IUPAC Name octadecylbenzene
Molecular Weight (g/mol) 330.60
PubChem CID 78187
Formula Weight 305.32
Percent Purity 99%
Chemical Name or Material Tri(4-morpholinyl)phosphine oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'