missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tri(2-methylphenyl)phosphine, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | (CH3C6H4)3P |
Quantity | 100 g |
Formula Weight | 304.37 |
Percent Purity | 97% |
Chemical Name or Material | Tri(2-methylphenyl)phosphine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'