Learn More
Thermo Scientific™ TREM2 Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding extracellular domain of human TREM2 (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus.
Brand: Thermo Scientific™ 11084H08H50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Expressed from DNA sequence encoding extracellular domain of human TREM2 (Met 1-Ser 174) fused to a polyhistidine tag at the C-terminus.
- Lyophilized in PBS, pH7.4, 5% mannitol, 5% trehalose, 0.02% Tween™-80
Specifications
Protein Structure Analysis, Immunogen, Protein Standard, Cell Biology and Analysis, Assay Standard | |
18.9kd | |
Human | |
<1 EU/μg | |
TREM2 | |
His Tag | |
Other proteins | |
≥85% by SDS-PAGE |
54209 | |
50μg | |
Truncated, Recombinant, Mutant | |
Triggering Receptor Expressed On Myeloid Cells 2 | |
TREM2 Recombinant Human Protein | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.