missing translation for 'onlineSavingsMsg'
Learn More

Trefoil Factor 3 Mouse anti-Mouse, Rat, eFluor 660, Clone: 5UCLNT3, eBioscience™

Mouse Monoclonal Antibody

£179.00

Specifications

Antigen Trefoil Factor 3
Clone 5UCLNT3
Host Species Murine
Gene Alias TFF3
Applications Immunohistochemistry, Microscopy
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15569946
View Documents
Affymetrix eBioscience
50-4758-82
100μg
£179.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment
07-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody 5UCLNT3 recognizes mouse and rat Trefoil Factor 3. Trefoil Factor 3 (TFF3) is a member of the trefoil family, which are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. TFF3 is a small peptide hormone secreted by mucus-producing cells and by epithelial cells of multiple tissues, including the gastrointestinal tract, kidney, and breast. TFF3 plays an essential role in both mucosal surface maintenance and restitution. TFF3 facilitates restoration of the intestinal epithelium as a protective barrier against injury by inhibiting apoptosis and promoting survival and migration of epithelial cells. TFF3 also plays a role in inducing airway epithelial ciliated cell differentiation. Levels of TFF3 change after renal injury and urinary levels can be used as a biomarker for kidney tubular injury. TFF3 may also play a role in breast tumor progression and tumor cell dissemination.
Specifications

Specifications

Trefoil Factor 3
Murine
Immunohistochemistry, Microscopy
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
5UCLNT3
TFF3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SDS

For Research Use Only.