missing translation for 'onlineSavingsMsg'
Learn More

Trefoil Factor 3, clone: 5UCLNT3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-4758-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15547446

  • £57.93 / 25µg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The monoclonal antibody 5UCLNT3 recognizes mouse and rat Trefoil Factor 3. Trefoil Factor 3 (TFF3) is a member of the trefoil family, which are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. TFF3 is a small peptide hormone secreted by mucus-producing cells and by epithelial cells of multiple tissues, including the gastrointestinal tract, kidney, and breast. TFF3 plays an essential role in both mucosal surface maintenance and restitution. TFF3 facilitates restoration of the intestinal epithelium as a protective barrier against injury by inhibiting apoptosis and promoting survival and migration of epithelial cells. TFF3 also plays a role in inducing airway epithelial ciliated cell differentiation. Levels of TFF3 change after renal injury and urinary levels can be used as a biomarker for kidney tubular injury. TFF3 may also play a role in breast tumor progression and tumor cell dissemination.Applications Reported: This 5UCLNT3 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy.Applications Tested: This 5UCLNT3 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded rat tissue using low pH antigen retrieval and can be used at less than or equal to 0.5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Trefoil Factor 3
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q62395, Q03191
ITF, P1B, TFI, polypeptide P1.B, trefoil factor 3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
MurineþRat
ImmunohistochemistryþMicroscopyþWestern Blot
5UCLNT3
Unconjugated
Tff3
Purified
Tff3
IgG1, kappa
25μg
Store at 2-8°C.
Primary
21786
Videos

For Research Use Only.