missing translation for 'onlineSavingsMsg'
Learn More

Trastuzumab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems

Mouse Monoclonal Antibody

£297.00 - £2160.00

Specifications

Antigen Trastuzumab
Classification Monoclonal
Conjugate Unconjugated
Host Species Human (HEK293)
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16498119
View Documents
ACROBiosystems
TRB-Y5b-100ug
100 μg
£297.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16408129
View Documents
ACROBiosystems
TRB-Y5b-1mg
1 mg
£2160.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trastuzumab Monoclonal antibody specifically detects Trastuzumab.
Specifications

Specifications

Trastuzumab
Unconjugated
RUO
-20°C
Monoclonal
Human (HEK293)
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.